1-(3-(dimethylamino)phenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89145-1G 89145-500MG 89145-5G
77473-700EA 497.49 USD
77473-700 77486-312 77496-928
1-(3-(dimethylamino)phenyl)cyclohexanol ≥95%
1-(3-(dimethylamino)phenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₄H₂₁NO
MW: 219.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23132067
CAS Number: 865374-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR