1-(4-Ethoxyphenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89130-1G 89130-500MG 89130-5G 89130-25G
77473-696EA 415.09 USD
77473-696 77486-308 77496-924 77505-874
1-(4-Ethoxyphenyl)cyclohexanol ≥97%
1-(4-Ethoxyphenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₄H₂₀O₂
MW: 220.307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222216
CAS Number: 1340394-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR