1-(3-Isopropylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89122-1G 89122-500MG 89122-5G 89122-25G
77473-692EA 370.8 USD
77473-692 77486-304 77496-920 77505-870
1-(3-Isopropylphenyl)cyclohexanol ≥95%
1-(3-Isopropylphenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD34771838
CAS Number: 676134-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR