1-(3-(tert-Butyl)phenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89117-1G 89117-500MG 89117-5G 89117-25G
77473-688EA 415.09 USD
77473-688 77486-300 77496-916 77505-866
1-(3-(tert-Butyl)phenyl)cyclohexanol ≥95%
1-(3-(tert-Butyl)phenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₆H₂₄O
MW: 232.361 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34817894
CAS Number: 94572-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR