1-(3-Methoxyphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89104-1G 89104-500MG 89104-5G 89104-25G
77473-680EA 346.08 USD
77473-680 77486-292 77496-910 77505-862
1-(3-Methoxyphenyl)cyclohexanol ≥95%
1-(3-Methoxyphenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701421
CAS Number: 1884-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR