Ethyl 2-(6-bromo-3-ethoxy-2-fluorophenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

89092-1G 89092-500MG 89092-5G 89092-25G
77473-676EA 415.09 USD
77473-676 77486-288 77496-906 77505-858
Ethyl 2-(6-bromo-3-ethoxy-2-fluorophenyl)-2-oxoacetate ≥97%
Ethyl 2-(6-bromo-3-ethoxy-2-fluorophenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₂H₁₂BrFO₄
MW: 319.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771826
CAS Number: 2918875-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR