1-(2-Isopropoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

88564-1G 88564-500MG 88564-5G
77473-672EA 538.69 USD
77473-672 77486-284 77496-902
1-(2-Isopropoxyphenyl)cyclobutanol ≥95%
1-(2-Isopropoxyphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23895252
CAS Number: 1521901-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR