9-propyl-9H-carbazole-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

88417-1G 88417-500MG 88417-5G 88417-25G
77473-668EA 415.09 USD
77473-668 77486-280 77496-898 77505-852
9-propyl-9H-carbazole-3-carbaldehyde ≥97%
9-propyl-9H-carbazole-3-carbaldehyde

Precaución: For research use only

Formula: C₁₆H₁₅NO
MW: 237.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00626048
CAS Number: 119607-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR