Ethyl 2-(2-Bromo-6-fluoro-4-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

87960-1G 87960-500MG 87960-5G 87960-25G
77473-664EA 415.09 USD
77473-664 77486-276 77496-894 77505-848
Ethyl 2-(2-Bromo-6-fluoro-4-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-Bromo-6-fluoro-4-methylphenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₁H₁₀BrFO₃
MW: 289.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771319
CAS Number: 2918889-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR