6-methylbenzo[d][1,3]dioxole-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

87618-1G 87618-500MG 87618-5G
77473-660EA 526.33 USD
77473-660 77486-272 77496-890
6-methylbenzo[d][1,3]dioxole-5-carboxylic acid ≥97%
6-methylbenzo[d][1,3]dioxole-5-carboxylic acid

Precaución: For research use only

Formula: C₉H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD00157046
CAS Number: 5025-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR