2-(3-Fluoro-4-(methylthio)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

87239-1G 87239-500MG 87239-5G 87239-25G
77473-656EA 170.98 USD
77473-656 77486-268 77496-884 77505-840
2-(3-Fluoro-4-(methylthio)phenyl)-1,3-dioxolane ≥97%
2-(3-Fluoro-4-(methylthio)phenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₁FO₂S
MW: 214.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771122
CAS Number: 2901082-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR