2-Methoxy-5-(methylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

87228-1G 87228-500MG 87228-5G 87228-25G
77473-652EA 415.09 USD
77473-652 77486-264 77496-880 77505-838
2-Methoxy-5-(methylthio)thiophene ≥95%
2-Methoxy-5-(methylthio)thiophene

Precaución: For research use only

Formula: C₆H₈OS₂
MW: 160.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00757496
CAS Number: 71908-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR