1-(5-Chlorothiophen-2-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

87222-1G 87222-500MG 87222-5G 87222-25G
77473-648EA 234.84 USD
77473-648 77486-260 77496-876 77505-834
1-(5-Chlorothiophen-2-yl)ethanol ≥97%
1-(5-Chlorothiophen-2-yl)ethanol

Precaución: For research use only

Formula: C₆H₇ClOS
MW: 162.637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443469
CAS Number: 125712-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR