Ethyl 2-(2,3-difluoro-6-(methoxymethoxy)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

87049-1G 87049-500MG 87049-5G 87049-25G
77473-636EA 415.09 USD
77473-636 77486-248 77496-868 77505-826
Ethyl 2-(2,3-difluoro-6-(methoxymethoxy)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,3-difluoro-6-(methoxymethoxy)phenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₂H₁₂F₂O₅
MW: 274.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771077
CAS Number: 2901099-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR