5-Bromo-1-fluoro-2-(methoxymethoxy)-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

86881-1G 86881-500MG 86881-5G 86881-25G
77473-632EA 415.09 USD
77473-632 77486-244 77496-864 77505-822
5-Bromo-1-fluoro-2-(methoxymethoxy)-3-methylbenzene ≥95%
5-Bromo-1-fluoro-2-(methoxymethoxy)-3-methylbenzene

Precaución: For research use only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770950
CAS Number: 2918846-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR