1-(3-Isopropoxy-5-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

86437-1G 86437-500MG 86437-5G
77473-628EA 518.09 USD
77473-628 77486-240 77496-860
1-(3-Isopropoxy-5-methylphenyl)ethanol ≥97%
1-(3-Isopropoxy-5-methylphenyl)ethanol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34770705
CAS Number: 2918892-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR