6-Fluoro-3-methoxy-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

86375-1G 86375-500MG 86375-5G 86375-25G
77473-620EA 415.09 USD
77473-620 77486-232 77496-852 77505-816
6-Fluoro-3-methoxy-2-(methylthio)phenol ≥97%
6-Fluoro-3-methoxy-2-(methylthio)phenol

Precaución: For research use only

Formula: C₈H₉FO₂S
MW: 188.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34817500
CAS Number: 2711285-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR