(2-Chloro-5-(trifluoromethyl)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86339-1G 86339-500MG 86339-5G 86339-25G
77473-616EA 347.11 USD
77473-616 77486-228 77496-848 77505-814
(2-Chloro-5-(trifluoromethyl)phenyl)trimethylsilane ≥95%
(2-Chloro-5-(trifluoromethyl)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₂ClF₃Si
MW: 252.736 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34687509
CAS Number: 148854-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR