(2-Fluoro-5-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86315-1G 86315-500MG 86315-5G 86315-25G
77473-612EA 397.58 USD
77473-612 77486-224 77496-844 77505-812
(2-Fluoro-5-methylphenyl)trimethylsilane ≥95%
(2-Fluoro-5-methylphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₅FSi
Storage Temperature: Ambient
MDL Number: MFCD34687508
CAS Number: 171089-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR