1-(3-Bromophenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

86282-1G 86282-500MG 86282-5G 86282-25G
77473-608EA 269.86 USD
77473-608 77486-220 77496-840 77505-808
1-(3-Bromophenyl)propan-1-ol ≥97%
1-(3-Bromophenyl)propan-1-ol

Precaución: For research use only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443305
CAS Number: 74157-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR