2-(2-Methoxy-6-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

86232-1G 86232-500MG 86232-5G 86232-25G
77473-604EA 415.09 USD
77473-604 77486-216 77496-836 77505-806
2-(2-Methoxy-6-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%
2-(2-Methoxy-6-(trifluoromethyl)phenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₁F₃O₃
MW: 248.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770661
CAS Number: 2901082-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR