(2-Ethylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86133-1G 86133-500MG 86133-5G 86133-25G
77473-600EA 339.9 USD
77473-600 77486-212 77496-832 77505-802
(2-Ethylphenyl)trimethylsilane ≥95%
(2-Ethylphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₁H₁₈Si
Storage Temperature: Ambient
MDL Number: MFCD11521270
CAS Number: 17988-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR