Trimethyl(4-phenoxyphenyl)silane ≥95%

Supplier: AOBChem USA

Warning

86118-1G 86118-500MG 86118-5G
77473-596EA 648.9 USD
77473-596 77486-208 77496-828
Trimethyl(4-phenoxyphenyl)silane ≥95%
Trimethyl(4-phenoxyphenyl)silane

Precaución: For research use only

Formula: C₁₅H₁₈OSi
Storage Temperature: Ambient
MDL Number: MFCD06201815
CAS Number: 17049-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR