1-(Benzo[d][1,3]dioxol-5-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85960-1G 85960-500MG 85960-5G
77473-592EA 676.71 USD
77473-592 77486-206 77496-824
1-(Benzo[d][1,3]dioxol-5-yl)cyclopentanol ≥95%
1-(Benzo[d][1,3]dioxol-5-yl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34770577
CAS Number: 95849-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR