1-(3-(dimethylamino)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85940-1G 85940-500MG 85940-5G
77473-588EA 497.49 USD
77473-588 77486-202 77496-820
1-(3-(dimethylamino)phenyl)cyclopentanol ≥95%
1-(3-(dimethylamino)phenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₃H₁₉NO
MW: 205.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34817473
CAS Number: 1518062-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR