1-(3-Fluoro-5-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85933-1G 85933-500MG 85933-5G 85933-25G
77473-584EA 415.09 USD
77473-584 77486-198 77496-816 77505-796
1-(3-Fluoro-5-methylphenyl)cyclopentanol ≥95%
1-(3-Fluoro-5-methylphenyl)cyclopentanol
Formula: C₁₂H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD23750728
CAS Number: 1541082-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR