1-(3-Fluoro-5-methoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85908-1G 85908-500MG 85908-5G 85908-25G
77473-580EA 226.6 USD
77473-580 77486-194 77496-812 77505-792
1-(3-Fluoro-5-methoxyphenyl)cyclopentanol ≥95%
1-(3-Fluoro-5-methoxyphenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₅FO₂
MW: 210.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770559
CAS Number: 2029519-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR