1-(4-(trimethylsilyl)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85901-1G 85901-500MG 85901-5G 85901-25G
77473-576EA 365.65 USD
77473-576 77486-190 77496-808 77505-788
1-(4-(trimethylsilyl)phenyl)cyclopentanol ≥95%
1-(4-(trimethylsilyl)phenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₄H₂₂OSi
Storage Temperature: Ambient
MDL Number: MFCD34770554
CAS Number: 2289319-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR