1-([1,1'-biphenyl]-4-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85889-1G 85889-500MG 85889-5G 85889-25G
77473-572EA 415.09 USD
77473-572 77486-186 77496-804 77505-784
1-([1,1'-biphenyl]-4-yl)cyclopentanol ≥95%
1-([1,1'-biphenyl]-4-yl)cyclopentanol

Precaución: For research use only

Formula: C₁₇H₁₈O
MW: 238.324 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20811149
CAS Number: 415898-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR