1-(4-(Methylthio)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85874-1G 85874-500MG 85874-5G 85874-25G
77473-568EA 399.64 USD
77473-568 77486-182 77496-800 77505-780
1-(4-(Methylthio)phenyl)cyclopentanol ≥95%
1-(4-(Methylthio)phenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₆OS
MW: 208.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30627909
CAS Number: 82955-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR