1-(5-Fluoro-2-methoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85860-1G 85860-500MG 85860-5G 85860-25G
77473-560EA 226.6 USD
77473-560 77486-174 77496-792 77505-774
1-(5-Fluoro-2-methoxyphenyl)cyclopentanol ≥95%
1-(5-Fluoro-2-methoxyphenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₅FO₂
MW: 210.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24305417
CAS Number: 1502214-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR