3-(1-hydroxycyclopentyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

85851-1G 85851-500MG 85851-5G
77473-556EA 486.16 USD
77473-556 77486-170 77496-788
3-(1-hydroxycyclopentyl)benzonitrile ≥97%
3-(1-hydroxycyclopentyl)benzonitrile

Precaución: For research use only

Formula: C₁₂H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD30477478
CAS Number: 2060050-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR