1-(3-Bromo-5-(methylthio)thiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

85846-1G 85846-500MG 85846-5G 85846-25G
77473-552EA 415.09 USD
77473-552 77486-166 77496-784 77505-770
1-(3-Bromo-5-(methylthio)thiophen-2-yl)ethanone ≥97%
1-(3-Bromo-5-(methylthio)thiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₇H₇BrOS₂
MW: 251.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770546
CAS Number: 2901102-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR