1-(3-Methoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85843-1G 85843-500MG 85843-5G 85843-25G
77473-548EA 284.28 USD
77473-548 77486-162 77496-780 77505-768
1-(3-Methoxyphenyl)cyclopentanol ≥95%
1-(3-Methoxyphenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD12152931
CAS Number: 142144-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR