1-(o-tolyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85837-1G 85837-500MG 85837-5G
77473-544EA 1135.06 USD
77473-544 77486-158 77496-776
1-(o-tolyl)cyclopentanol ≥95%
1-(o-tolyl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD12152936
CAS Number: 100256-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR