1-(3-Chloro-4-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85811-1G 85811-500MG 85811-5G 85811-25G
77473-540EA 394.49 USD
77473-540 77486-154 77496-772 77505-766
1-(3-Chloro-4-methylphenyl)cyclobutanol ≥95%
1-(3-Chloro-4-methylphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₁H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD30156731
CAS Number: 1697811-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR