1-(4-(Methylthio)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85767-1G 85767-500MG 85767-5G
77473-536EA 486.16 USD
77473-536 77486-150 77496-768
1-(4-(Methylthio)phenyl)cyclobutanol ≥95%
1-(4-(Methylthio)phenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₁H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD30627904
CAS Number: 29480-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR