1-(3,5-dimethoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85747-1G 85747-500MG 85747-5G
77473-532EA 486.16 USD
77473-532 77486-146 77496-764
1-(3,5-dimethoxyphenyl)cyclobutanol ≥95%
1-(3,5-dimethoxyphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23827964
CAS Number: 1520873-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR