1-(2-Methoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85722-1G 85722-500MG 85722-5G 85722-25G
77473-520EA 255.44 USD
77473-520 77486-134 77496-752 77505-756
1-(2-Methoxyphenyl)cyclobutanol ≥95%
1-(2-Methoxyphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD21788692
CAS Number: 1508180-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR