1-(3,4-Dichlorophenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

85661-1G 85661-500MG 85661-5G 85661-25G
77473-512EA 286.34 USD
77473-512 77486-126 77496-744 77505-752
1-(3,4-Dichlorophenyl)-2-fluoroethanone ≥95%
1-(3,4-Dichlorophenyl)-2-fluoroethanone

Precaución: For research use only

Formula: C₈H₅Cl₂FO
MW: 207.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28970126
CAS Number: 1219632-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR