ω-Fluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-1-phenylethanone, Phenacyl fluoride

85637-1G 85637-5G 85637-25G
77473-508EA 60.77 USD
77473-508 77496-740 77505-750
ω-Fluoroacetophenone ≥97%
ω-Fluoroacetophenone

Precaución: For research use only

Formula: C₈H₇FO
MW: 138.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08461612
CAS Number: 450-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR