5-Ethylthiophene-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

85324-1G 85324-500MG 85324-5G 85324-25G
77473-504EA 187.46 USD
77473-504 77486-120 77496-736 77505-746
5-Ethylthiophene-3-carboxylic acid ≥97%
5-Ethylthiophene-3-carboxylic acid

Precaución: For research use only

Formula: C₇H₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD03419849
CAS Number: 19156-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR