2-(3-Bromo-2-fluoro-6-methoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

85012-1G 85012-500MG 85012-5G 85012-25G
77473-492EA 415.09 USD
77473-492 77486-108 77496-726 77505-736
2-(3-Bromo-2-fluoro-6-methoxyphenyl)-1,3-dioxolane ≥95%
2-(3-Bromo-2-fluoro-6-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770307
CAS Number: 2901109-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR