2-(3-Bromo-6-chloro-2-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

84925-1G 84925-500MG 84925-5G 84925-25G
77473-488EA 415.09 USD
77473-488 77486-104 77496-720 77505-730
2-(3-Bromo-6-chloro-2-methoxyphenyl)-1,3-dioxolane ≥97%
2-(3-Bromo-6-chloro-2-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrClO₃
MW: 293.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770258
CAS Number: 2901099-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR