3,4,5-tribromobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

84614-1G 84614-500MG 84614-5G 84614-25G
77473-480EA 135.96 USD
77473-480 77486-096 77496-712 77505-722
3,4,5-tribromobenzoic acid ≥95%
3,4,5-tribromobenzoic acid

Precaución: For research use only

Formula: C₇H₃Br₃O₂
MW: 358.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075794
CAS Number: 618-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR