2-Amino-5-bromo-3-fluorobenzonitrile ≥97%
Supplier: AOBChem USA
|
Warning
|
84182-1G
84182-500MG
84182-5G
84182-25G
77473-476EA
72.1
USD
77473-476
77486-092
77496-706
77505-716
2-Amino-5-bromo-3-fluorobenzonitrile ≥97%
2-Amino-5-bromo-3-fluorobenzonitrile
Precaución: For research use only
Formula:
C₇H₄BrFN₂ MW: 215.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17011768 CAS Number: 1209498-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...