1-Butyl-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

83910-1G 83910-500MG 83910-5G 83910-25G
77473-472EA 397.58 USD
77473-472 77486-088 77496-702 77505-712
1-Butyl-4-methoxybenzene ≥95%
1-Butyl-4-methoxybenzene

Precaución: For research use only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD02259755
CAS Number: 18272-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR