1-(3-Bromo-5-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

83815-1G 83815-500MG 83815-5G 83815-25G
77473-468EA 297.67 USD
77473-468 77486-084 77496-698 77505-708
1-(3-Bromo-5-methylphenyl)cyclopentanol ≥95%
1-(3-Bromo-5-methylphenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₅BrO
MW: 255.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769652
CAS Number: 2766483-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR