(4-(1,1-difluoroethyl)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

83655-1G 83655-500MG 83655-5G 83655-25G
77473-464EA 151.41 USD
77473-464 77486-080 77496-694 77505-704
(4-(1,1-difluoroethyl)phenyl)boronic acid ≥95%
(4-(1,1-difluoroethyl)phenyl)boronic acid

Precaución: For research use only

Formula: C₈H₉BF₂O₂
MW: 185.964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26404335
CAS Number: 1162257-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR