(2-Hydroxy-3-(methylthio)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

83425-1G 83425-500MG 83425-5G 83425-25G
77473-460EA 415.09 USD
77473-460 77486-076 77496-692 77505-702
(2-Hydroxy-3-(methylthio)phenyl)boronic acid ≥97%
(2-Hydroxy-3-(methylthio)phenyl)boronic acid

Precaución: For research use only

Formula: C₇H₉BO₃S
Storage Temperature: Ambient
MDL Number: MFCD22414770
CAS Number: 916673-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR